Learn More
Potassium phthalimide, 98+%, Thermo Scientific Chemicals
33.90 € - 354.00 €
Chemical Identifiers
CAS | 1074-82-4 |
---|---|
Molecular Formula | C8H4KNO2 |
Molecular Weight (g/mol) | 185.22 |
MDL Number | MFCD00005887 |
InChI Key | FYRHIOVKTDQVFC-UHFFFAOYSA-M |
Synonym | potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil |
PubChem CID | 3356745 |
IUPAC Name | potassium;isoindol-2-ide-1,3-dione |
SMILES | [K+].O=C1[N-]C(=O)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439033
|
Thermo Scientific Alfa Aesar
A11134.22 |
100 g |
33.90 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449033
|
Thermo Scientific Alfa Aesar
A11134.36 |
500 g |
90.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459033
|
Thermo Scientific Alfa Aesar
A11134.0E |
2500 g |
354.00 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium phthalimide is used as an intermediate in the synthesis of N-alkylated phthalimides, which is involved in the preparation of primary amines (Gabriel synthesis) by the hydrolysis reaction. It is also used as an intermediate for synthetic indigo, pigments, dyes and pharmaceuticals. Further, it is employed as an organocatalyst for the cyanosilylation of various carbonyl compounds under extremely mild conditions. In addition to this, it serves as a reagent for the transformation of allyl- and alkyl halides into protected primary amines.
Solubility
Soluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
1074-82-4 | |
185.22 | |
FYRHIOVKTDQVFC-UHFFFAOYSA-M | |
3356745 | |
[K+].O=C1[N-]C(=O)C2=CC=CC=C12 |
C8H4KNO2 | |
MFCD00005887 | |
potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil | |
potassium;isoindol-2-ide-1,3-dione |
Specifications
1074-82-4 | |
1.63 | |
MFCD00005887 | |
Moisture sensitive | |
Soluble in water. | |
[K+].O=C1[N-]C(=O)C2=CC=CC=C12 | |
185.22 | |
185.23 | |
Potassium phthalimide |
>300°C | |
C8H4KNO2 | |
3598719 | |
potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil | |
FYRHIOVKTDQVFC-UHFFFAOYSA-M | |
potassium;isoindol-2-ide-1,3-dione | |
3356745 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 214-046-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only