missing translation for 'onlineSavingsMsg'
Learn More

Potassium phosphate monobasic, puriss. p.a., ACS Reagent, Honeywell Fluka™

Puriss. p.a., Buffer substance, Anhydrous, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99.5-100.5%

70.04 € - 533.17 €

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium;dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15625940
View Documents
Honeywell-Fluka
60220-500G
500 g
70.04 €
500g
Please to purchase this item. Need a web account? Register with us today!
15605940
View Documents
Honeywell-Fluka
60220-1KG
1 kg
154.79 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15615940
View Documents
Honeywell-Fluka
60220-2.5KG
2.5 kg
243.91 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
15635940
View Documents
Honeywell-Fluka
60220-5KG
5 kg
533.17 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium;dihydrogen phosphate
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
[K+].OP(O)([O-])=O

Specifications

7778-77-0
KH2PO4
NONH for all modes of transport
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium;dihydrogen phosphate
516951
136.09g/mol
ACS/puriss. p.a.
Potassium phosphate monobasic
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
99.5 to 100.5%
2.338g/mL at 25°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TC6615500

Product Identifier
  • Potassium phosphate monobasic

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.