missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium permanganate solution, Volumetric, 0.2 M KMnO4 (1N), Honeywell Fluka™
Volumetric, 0.2 M KMnO4 (1N)
64.89 € - 337.82 €
Chemical Identifiers
CAS | 7722-64-7 |
---|---|
Molecular Formula | KMnO4 |
Molecular Weight (g/mol) | 158.032 |
MDL Number | MFCD00011364 |
InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
PubChem CID | 516875 |
IUPAC Name | potassium;permanganate |
SMILES | [O-][Mn](=O)(=O)=O.[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15644840
|
Honeywell-Fluka
35184-1L |
1 L |
64.89 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654840
|
Honeywell-Fluka
35184-6X1L |
6 x 1 L |
337.82 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7722-64-7 | |
158.032 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
516875 | |
[O-][Mn](=O)(=O)=O.[K+] |
KMnO4 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
potassium;permanganate |
Specifications
7722-64-7 | |
MFCD00011364 | |
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
[O-][Mn](=O)(=O)=O.[K+] | |
158.032 | |
158.03g/mol | |
Potassium permanganate solution |
KMnO4 | |
UN3082 | |
VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
potassium;permanganate | |
516875 | |
Glass bottle |
Safety and Handling
P273-P501
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- potassium permanganate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : potassium permanganate