missing translation for 'onlineSavingsMsg'
Learn More

Potassium Permanganate Solution, EU Pharmacopoeia Grade, 0.02M, Fisher Chemical™

Product Code. 12943594
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.02mol/L
Product Identifier
  • Potassium permanganate 0.02M Volumetric Solution
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'