Learn More
Potassium osmate(VI) dihydrate, 51.0-52.0% Os, Thermo Scientific Chemicals
101.51 € - 347.73 €
Chemical Identifiers
CAS | 10022-66-9 |
---|---|
Molecular Formula | K2O4Os·2H2O |
Molecular Weight (g/mol) | 368.42 |
MDL Number | MFCD00149919 |
InChI Key | DGODWNOPHMXOTR-UHFFFAOYSA-N |
Synonym | potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate |
PubChem CID | 53393272 |
IUPAC Name | dipotassium;dioxido(dioxo)osmium;dihydrate |
SMILES | O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10194210
|
Thermo Scientific Acros
197665000 |
500 mg | Glass bottle |
101.51 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10368670
|
Thermo Scientific Acros
197660025 |
2.5 g | Glass bottle |
347.73 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10022-66-9 | |
368.42 | |
DGODWNOPHMXOTR-UHFFFAOYSA-N | |
53393272 | |
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] |
K2O4Os·2H2O | |
MFCD00149919 | |
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate | |
dipotassium;dioxido(dioxo)osmium;dihydrate |
Specifications
10022-66-9 | |
Crystalline Powder | |
K2OsO4·2H2O | |
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate | |
DGODWNOPHMXOTR-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)osmium;dihydrate | |
53393272 | |
51.0 to 52% Os |
Violet | |
K2O4Os·2H2O | |
MFCD00149919 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether | |
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+] | |
368.42 | |
368.42 | |
Potassium osmate(VI) dihydrate, 51.0-52.0% Os |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May intensify fire; oxidizer.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 243-247-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only