missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium nitrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%
Chemical Identifiers
CAS | 7757-79-1 |
---|---|
Molecular Formula | KNO3 |
Molecular Weight (g/mol) | 101.10 |
MDL Number | MFCD00011409 |
InChI Key | FGIUAXJPYTZDNR-UHFFFAOYSA-N |
Synonym | potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras |
PubChem CID | 24434 |
ChEBI | CHEBI:63043 |
SMILES | [K+].[O-][N+]([O-])=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7757-79-1 | |
101.10 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
24434 | |
[K+].[O-][N+]([O-])=O |
KNO3 | |
MFCD00011409 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
CHEBI:63043 |
Specifications
7757-79-1 | |
MFCD00011409 | |
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
CHEBI:63043 | |
≥99% | |
Potassium Nitrate, Reag. Ph. Eur. reag. ISO |
KNO3 | |
1486 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium;nitrate | |
24434 | |
101.1g/mol | |
ACS/puriss. p.a. |
Safety and Handling
P220-P273
H272-H412
missing translation for 'einecsNumber' : 231-818-8
missing translation for 'rtecsNumber' : TT3700000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Potassium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.