missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide Solution, EU Pharmacopoeia Grade, 0.1M, Fisher Chemical™

Product Code. 12903594
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.1mol/L
compliance-icons
Product Identifier
  • Potassium iodide 0.1M Volumetric Solution
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P314-Get medical advice/attention if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : potassium iodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'