missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 1 kg |
Linear Formula | KIO3 |
UN Number | 1479 |
Formula Weight | 214g/mol |
Percent Purity | 99.7 to 104% |
Grade | puriss. p.a. |
Density | 3.93g/mL at 25°C |
Chemical Name or Material | Potassium Iodate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction