missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodate, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10562354
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-05-6, 7732-18-5
Molecular Formula IKO3
Molecular Weight (g/mol) 214
MDL Number MFCD00011406
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

Melting Point 560°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula KIO3
Merck Index 15, 7763
Solubility Information Solubility in water: soluble
Formula Weight 214
Percent Purity 99.50%
Grade Analytical
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 3.93
Density 3.9300g/mL
Chemical Name or Material Potassium iodate, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.