missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, Volumetric, 0.1 M KOH in ethanol (0.1N), Honeywell Fluka™
Volumetric, 0.1 M KOH in ethanol (0.1N)
89.76 €
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
IUPAC Name | potassium;hydroxide |
SMILES | [OH-].[K+] |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
CHEBI:32035 | |
[OH-].[K+] |
HKO | |
MFCD00003553 | |
14797 | |
potassium;hydroxide |
Specifications
1310-58-3 | |
KOH | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
14797 | |
56.11g/mol |
HKO | |
0.1M in ethanol | |
UN1170 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
14°C (57.2°F) | |
CHEBI:32035 | |
Potassium hydroxide solution |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Potassium hydroxide solution
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanol; ethyl alcohol, Toluene, potassium hydroxide; caustic potash, hydrazine
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title