missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 1.0M, Honeywell Fluka™

Product Code. 15633560
Click to view available options
Quantity:
500 mL
2 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Volumetric, 1.0 M KOH (1.0 N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
SMILES [OH-].[K+]

Specifications

Linear Formula KOH
Concentration 1.0M
UN Number 1814
Density 1.046g/mL at 25°C
Quantity 2.5 L
IUPAC Name potassium hydroxide
Formula Weight 56.11g/mol
Chemical Name or Material Potassium Hydroxide Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'