missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hydroxide solution, Volumetric, 0.5 M KOH in ethanol (0.5N), Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Linear Formula | KOH |
UN Number | UN2924 |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
Quantity | 6 x 500 mL |
IUPAC Name | potassium hydroxide |
Flash Point | 11°C (51.8°F) |
Formula Weight | 56.11 |
Chemical Name or Material | Potassium hydroxide solution, volumetric, 0.5 M KOH in ethanol (0.5N) |
Safety and Handling
Product Identifier
- Potassium hydroxide solution
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hydroxide;caustic potash, Toluene, ethanol; ethylalcohol, Hydrazinehydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction