missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydroxide solution, 0.1M in isopropanol, Honeywell™

Product Code. 15634780
Click to view available options
Quantity:
1 L

Specifications

CAS 1310-58-3
Molecular Formula HKO
Linear Formula KOH
Concentration 0.1M in isopropanol
MDL Number MFCD00003553
UN Number UN1219
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
Flash Point 12°C (53.6°F)
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Chemical Name or Material Potassium hydroxide solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.