missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
27.50 € - 105.00 €
Chemical Identifiers
CAS | 1310-58-3 |
---|---|
Molecular Formula | HKO |
Molecular Weight (g/mol) | 56.11 |
MDL Number | MFCD00003553 |
InChI Key | KWYUFKZDYYNOTN-UHFFFAOYSA-M |
Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 |
PubChem CID | 14797 |
ChEBI | CHEBI:32035 |
SMILES | [OH-].[K+] |
Description
Chemical Identifiers
1310-58-3 | |
56.11 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
14797 | |
[OH-].[K+] |
HKO | |
MFCD00003553 | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
CHEBI:32035 |
Specifications
13 | |
HKO | |
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693 | |
[OH-].[K+] | |
56.11 | |
CHEBI:32035 | |
Ampoule |
1310-58-3 | |
MFCD00003553 | |
KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
potassium;hydroxide | |
14797 | |
56.11g/mol | |
Liquid |