missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, Certified AR for Analysis, Fisher Chemical™

38.80 € - 95.50 €

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
SMILES [K+].OS([O-])(=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10408420
View Documents
Fisher Chemical
P/5440/50
250 g
38.80 €
250g
Please to purchase this item. Need a web account? Register with us today!
10408610
View Documents
Fisher Chemical
P/5440/53
500 g
64.30 €
500g
Please to purchase this item. Need a web account? Register with us today!
10418610
View Documents
Fisher Chemical
P/5440/60
1 kg
95.50 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Potassium Hydrogen Sulfate, Certified AR for Analysis

Specifications

Chemical Identifiers

7646-93-7
136.16
CHKVPAROMQMJNQ-UHFFFAOYSA-M
516920
HKO4S
MFCD00011404
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
[K+].OS([O-])(=O)=O

Specifications

214°C
White
Solid
MFCD00011404
CHKVPAROMQMJNQ-UHFFFAOYSA-M
potassium hydrogen sulfate
516920
Plastic powder jar
7646-93-7
1
HKO4S
potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
[K+].OS([O-])(=O)=O
136.16
136.16g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium bisulfate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.