missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, ACS reagent, acidimetric standard, Thermo Scientific Chemicals

42.61 € - 328.64 €

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.22
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10712412
View Documents
Thermo Scientific Acros
424061000
100 g Plastic bottle
42.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
10351211
View Documents
Thermo Scientific Acros
424065000
500 g Plastic bottle
113.57 €
500g
Please to purchase this item. Need a web account? Register with us today!
10447493
View Documents
Thermo Scientific Acros
424060025
2.5 kg Plastic bottle
328.64 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

877-24-7
204.22
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
MFCD00013070
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
potassium;2-carboxybenzoate

Specifications

877-24-7
White
Authentic
2-(HO2C)C6H4CO2K
09, 791
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
99.95 to 100.05% (on dried substance)
0.005% max
Potassium hydrogen phthalate
295.0°C to 300.0°C
4.00 to 4.02 (0.05M soln. at 25°C ±0.2°C)
C8H5KO4
MFCD00013070
15, 7733
Solubility in water: 80g/L (20°C). Other solubilities: 330g/L water (100°C),slightly soluble in alcohol
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.22
204.22
ACS Reagent
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 212-889-4

missing translation for 'rtecsNumber' : CZ4326000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.