missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen diiodate, 99.85%, extra pure, Thermo Scientific Chemicals

96.50 € - 323.00 €

Chemical Identifiers

CAS 13455-24-8
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.9
MDL Number MFCD00011400
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10401451
View Documents
Thermo Scientific Acros
211471000
100 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10022690
View Documents
Thermo Scientific Acros
211475000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

13455-24-8
389.9
ACAYDTMSDROWHW-UHFFFAOYSA-M
23700942
OI(=O)=O.[O-]I(=O)=O.[K+]
HI2KO6
MFCD00011400
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
potassium;iodic acid;iodate

Specifications

13455-24-8
Crystals or Powder
HI2KO6
MFCD00011400
Solubility in water: 13g/L (15°C)
OI(=O)=O.[O-]I(=O)=O.[K+]
389.9
389.9
Extra Pure
White to Yellow
99.8% min. (Iodometry)
KH(IO3)2
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
23700942
99.85%
Potassium hydrogen diiodate, p.a.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
IF SWALLOWED: Ri

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 236-650-9

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only