missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Carbonate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10204210
Click to view available options
Quantity:
500 g
1 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Chemische Identifikatoren

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
SMILES [K+].OC([O-])=O

Spezifikation

Melting Point 292°C
Color Yellow
pH 8.2
Physical Form Solid
Quantity 1 kg
IUPAC Name potassium;hydrogen carbonate
Formula Weight 100.12g/mol
Packaging Plastic powder jar
Produktidentifikator
  • Potassium hydrogen carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'