Learn More
Potassium hexachloroplatinate(IV), Pt 39.6%, Thermo Scientific Chemicals
41.80 € - 403.00 €
Chemical Identifiers
CAS | 16921-30-5 |
---|---|
Molecular Formula | Cl6K2Pt |
Molecular Weight (g/mol) | 485.98 |
MDL Number | MFCD00011389 |
InChI Key | DPAIVKJGTXERIM-UHFFFAOYSA-H |
Synonym | potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride |
PubChem CID | 61856 |
IUPAC Name | dipotassium;hexachloroplatinum(2-) |
SMILES | [K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11346548
|
Thermo Scientific Alfa Aesar
012169.01 |
0.25 g |
41.80 €
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336548
|
Thermo Scientific Alfa Aesar
012169.03 |
1 g |
121.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326548
|
Thermo Scientific Alfa Aesar
012169.06 |
5 g |
403.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexachloroplatinate(IV) is used for the determination of potassium by gravimetric analysis. It is also used to prepare potassium tetrchloroplatinate by reduction using hydrazine dihydrochloride. Further, it is used in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16921-30-5 | |
485.98 | |
DPAIVKJGTXERIM-UHFFFAOYSA-H | |
61856 | |
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl |
Cl6K2Pt | |
MFCD00011389 | |
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride | |
dipotassium;hexachloroplatinum(2-) |
Specifications
250°C (decomposition) | |
Yellow | |
Cl6K2Pt | |
MFCD00011389 | |
14,7636 | |
Soluble in hot water; Slightly soluble in cold water; Practically insoluble in alcoholSoluble in hot water; Slightly soluble in cold water; Insoluble in alcohol. | |
[K+].[K+].Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl | |
485.98 | |
486.01 | |
Odorless | |
Hygroscopic | |
Potassium hexachloroplatinate(IV) |
16921-30-5 | |
Crystalline Powder | |
K2PtCl6 | |
UN3288 | |
potassium hexachloroplatinate iv, potassium platinic chloride, potassium hexachloroplatinate, dipotassium platinum hexachloride, platinic potassium chloride, dipotassium hexachloroplatinate, potassium platinum chloride, dipotassium hexachloroplatinate 2-, ccris 7824, platinum potassium chloride | |
DPAIVKJGTXERIM-UHFFFAOYSA-H | |
dipotassium;hexachloroplatinum(2-) | |
61856 | |
Pt 39.6% | |
≥1.825 | |
3.50 g/mL |
Safety and Handling
GHS H Statement
H301-H334-H372-H318-H290-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
P234-P260-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P314-P330-P333+P313-P342+P311-P363-P390-P501c
H290-H301-H317-H318-H334-H335-H373
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 240-979-3
missing translation for 'rtecsNumber' : TP1650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only