Learn More
Potassium hexachloroosmate(IV), min 39% Os, Thermo Scientific Chemicals
217.08 € - 750.74 €
Chemical Identifiers
CAS | 16871-60-6 |
---|---|
Molecular Formula | Cl6K2Os |
Molecular Weight (g/mol) | 481.11 |
MDL Number | MFCD00011371 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10348670
|
Thermo Scientific Acros
197650010 |
1 g | Glass bottle |
217.08 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358670
|
Thermo Scientific Acros
197650050 |
5 g | Glass bottle |
750.74 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16871-60-6 | |
Cl6K2Os | |
481.11 | |
MFCD00011371 |
Specifications
600.0°C | |
Bordeaux to Purple | |
Cl6K2Os | |
MFCD00011371 | |
Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol | |
481.11 | |
3.42 |
16871-60-6 | |
Powder | |
K2OsCl6 | |
15, 7756 | |
481.11 | |
≥39% (Os) | |
Potassium hexachloroosmate(IV), min 39% Os |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 240-893-6
missing translation for 'tsca' : TSCA
- Potassium hexachloroosmate(IV)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.