Learn More
Potassium hexachloroiridate(IV), Ir 39% min, Thermo Scientific Chemicals
314.00 € - 5804.00 €
Chemical Identifiers
CAS | 16920-56-2 |
---|---|
Molecular Formula | Cl6IrK2 |
Molecular Weight (g/mol) | 483.11 |
MDL Number | MFCD00011369 |
InChI Key | SCPDBYGTLSSBQR-UHFFFAOYSA-N |
Synonym | potassium hexachloroiridate iv, dipotassium hexachloroiridate, cl6ir.2k, dipotassium hexachloroiridate 2- |
PubChem CID | 11049163 |
IUPAC Name | dipotassium;hexachloroiridium(2-) |
SMILES | [Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[K+].[K+].[Ir] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391978
|
Thermo Scientific Alfa Aesar
011888.03 |
1 g |
314.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11381978
|
Thermo Scientific Alfa Aesar
011888.06 |
5 g |
1479.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371978
|
Thermo Scientific Alfa Aesar
011888.14 |
25 g |
5804.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium hexachloroiridate(IV) is used to prepare platinum nanoparticles (PNPs) by reduction with trisodium citrate and sodium dodecyl sulfate as stabilizer reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16920-56-2 | |
483.11 | |
SCPDBYGTLSSBQR-UHFFFAOYSA-N | |
11049163 | |
[Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[K+].[K+].[Ir] |
Cl6IrK2 | |
MFCD00011369 | |
potassium hexachloroiridate iv, dipotassium hexachloroiridate, cl6ir.2k, dipotassium hexachloroiridate 2- | |
dipotassium;hexachloroiridium(2-) |
Specifications
(decomposition) | |
Crystalline | |
MFCD00011369 | |
Insoluble in water. | |
[Cl].[Cl].[Cl].[Cl].[Cl].[Cl].[K+].[K+].[Ir] | |
483.11 | |
483.12 | |
Odorless | |
Potassium hexachloroiridate(IV) |
16920-56-2 | |
Cl6IrK2 | |
potassium hexachloroiridate iv, dipotassium hexachloroiridate, cl6ir.2k, dipotassium hexachloroiridate 2- | |
SCPDBYGTLSSBQR-UHFFFAOYSA-N | |
dipotassium;hexachloroiridium(2-) | |
11049163 | |
Ir 39% min. | |
Hygroscopic |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 240-976-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only