missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexabromopalladate(IV), Premion™, 99.999% (metals basis), Pd 15.5% min, Thermo Scientific Chemicals

102.51 € - 422.10 €

Chemical Identifiers

CAS 16919-74-7
Molecular Formula Br6K2Pd
Molecular Weight (g/mol) 664.041
MDL Number MFCD03093651
InChI Key SKCNWISEAGYZFC-UHFFFAOYSA-H
Synonym potassium hexabromopalladate iv , premion
PubChem CID 121233751
IUPAC Name dipotassium;tetrabromopalladium;dibromide
SMILES [K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314998
View Documents
Thermo Scientific Alfa Aesar
043947.03
1 g
102.51 €
1g
Please to purchase this item. Need a web account? Register with us today!
11324998
View Documents
Thermo Scientific Alfa Aesar
043947.06
5 g
422.10 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16919-74-7
664.041
SKCNWISEAGYZFC-UHFFFAOYSA-H
121233751
[K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
Br6K2Pd
MFCD03093651
potassium hexabromopalladate iv , premion
dipotassium;tetrabromopalladium;dibromide

Specifications

16919-74-7
Br6K2Pd
potassium hexabromopalladate iv , premion
[K+].[K+].[Br-].[Br-].Br[Pd](Br)(Br)Br
664.041
664.04
Premion™
(metals basis)
Crystalline
MFCD03093651
SKCNWISEAGYZFC-UHFFFAOYSA-H
dipotassium;tetrabromopalladium;dibromide
121233751
99.999%
Pd 15.5% min
Potassium hexabromopalladate (IV)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.