Learn More
Potassium ferrocyanide trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
55.40 € - 189.00 €
Chemical Identifiers
CAS | 14459-95-1 |
---|---|
Molecular Formula | C6H6FeK4N6O3 |
Molecular Weight (g/mol) | 422.39 |
MDL Number | MFCD00167023 |
InChI Key | NLYKFUFIUWPPIZ-UHFFFAOYSA-N |
Synonym | Potassium hexacyanoferrate(II) |
IUPAC Name | tetrapotassium hexacyanoironbis(ylium) trihydrate |
SMILES | O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10216460
|
Thermo Scientific Acros
211095000 |
500 g | Plastic bottle |
55.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10504474
|
Thermo Scientific Acros
211090025 |
2.5 kg | Plastic bottle |
189.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14459-95-1 | |
422.39 | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
tetrapotassium hexacyanoironbis(ylium) trihydrate |
C6H6FeK4N6O3 | |
MFCD00167023 | |
Potassium hexacyanoferrate(II) | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N |
Specifications
70.0°C | |
Yellow | |
99+% | |
K4Fe(CN)6·3H2O | |
15, 7752 | |
Solubility in water: 270g/L (12°C). Other solubilities: insoluble in alcohol | |
O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N | |
422.39 | |
≥99% | |
1.85 | |
Potassium ferrocyanide trihydrate, p.a. |
14459-95-1 | |
Crystalline Powder | |
C6H6FeK4N6O3 | |
MFCD00167023 | |
Potassium hexacyanoferrate(II) | |
NLYKFUFIUWPPIZ-UHFFFAOYSA-N | |
tetrapotassium hexacyanoironbis(ylium) trihydrate | |
422.4 | |
Analytical | |
1.8500g/mL |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
- Potassium ferrocyanide trihydrate
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : Potassium ferrocyanide trihydrate, Ferrate(4-), hexakis(cyano-C)-, tetrapotassium, (OC-6-11)-
RUO – Research Use Only