missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferricyanide, 98%, pure, Thermo Scientific Chemicals

Product Code. 10636974
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange to Red
Physical Form Crystalline Powder and/or Crystals
Quantity 5 kg
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula K3Fe(CN)6
Fieser 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
Merck Index 15, 7751
Solubility Information Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name tripotassium hexacyanoirontris(ylium)
Formula Weight 329.26
Percent Purity 98%
Grade Pure
Packaging Plastic Bottle
Density 1.85
Chemical Name or Material Potassium ferricyanide, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.