missing translation for 'onlineSavingsMsg'
Learn More

Potassium dihydrogen phosphate, BAKER, J.T. Baker™

Product Code. 10542391
Click to view available options
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Boiling Point ∼100°C (1013 hPa)
Melting Point 253°C
CAS 7778-77-0
Quantity 1 kg
Assay Percent Range min. 99%
Molecular Formula H2KO4P
MDL Number MFCD00011401 MFCD00147253
Synonym Potassium dihydrogen orthophosphate , Potassium phosphate monobasic , Monobasic Potassium Phosphate
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES [K+].OP(O)([O-])=O
IUPAC Name potassium dihydrogen phosphate
Molecular Weight (g/mol) 136.08
Loss on Drying max. 0.1% (at 105°C)
Packaging Plastic bottle for solids
Density 2.3 g/cm³ (20°C)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.