missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Crystal, Primary Standard, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 14874641
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 7778-50-9
Physical Form Crystals
Quantity 500 g
Molecular Formula Cr2K2O7
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 294.182
PubChem CID 24502
ChEBI CHEBI:53444
Formula Weight 294.19
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.68kg
Chemical Name or Material Potassium Dichromate, Primary Standard
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'