missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, Honeywell™

Product Code. 15665930
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Purum p.a.,≥97.0% (AT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Quantity 250 g
Linear Formula KCN
UN Number UN1680
Beilstein 3593645
Formula Weight 65.12g/mol
Percent Purity ≥97% (AT)
Chemical Name or Material Potassium cyanide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.