Learn More
Potassium cyanate, 97%, pure, Thermo Scientific Chemicals
23.62 € - 89.95 €
Chemical Identifiers
CAS | 590-28-3 |
---|---|
Molecular Formula | CKNO |
Molecular Weight (g/mol) | 81.12 |
MDL Number | MFCD00011411 |
InChI Key | GKKCIDNWFBPDBW-UHFFFAOYSA-M |
Synonym | potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 |
PubChem CID | 11378442 |
ChEBI | CHEBI:38904 |
IUPAC Name | potassium;cyanate |
SMILES | [K+].[O-]C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10289403
|
Thermo Scientific Acros
201111000 |
100 g | Plastic bottle |
23.62 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10430861
|
Thermo Scientific Acros
201115000 |
500 g | Plastic bottle |
54.57 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10720711
|
Thermo Scientific Acros
201110010 |
1 kg | Plastic bottle |
89.95 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-28-3 | |
81.12 | |
GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
11378442 | |
potassium;cyanate |
CKNO | |
MFCD00011411 | |
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
CHEBI:38904 | |
[K+].[O-]C#N |
Specifications
315.0°C | |
White | |
97% | |
KOCN | |
01,1059 | |
potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
potassium;cyanate | |
11378442 | |
81.12 | |
Pure |
590-28-3 | |
Crystalline Powder and/or Chunks | |
CKNO | |
MFCD00011411 | |
15, 7746 | |
Solubility in water: 750g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[O-]C#N | |
81.12 | |
CHEBI:38904 | |
97% | |
Potassium cyanate, Pure |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-676-3
- Potassium cyanate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.