missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. 15623330
Click to view available options
Quantity:
250 g
1 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Boiling Point 1,000°C
Melting Point 971°C (literature)
CAS Max % 100.0000%
Quantity 50 kg
Linear Formula Al(NO3)3 · 9H2O
UN Number UN3288
IUPAC Name dipotassium;dioxido(dioxo)chromium
Formula Weight 194.19
Percent Purity >99.5%
Grade ACS Reagent/Reagent Ph. Eur.
Packaging Fiberboard Box
Density 2.732 g/cm3; Vapor Density: 6.7 (vs air)
Chemical Name or Material Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.