Learn More
Potassium chromate, ACS reagent, Thermo Scientific Chemicals
33.40 € - 106.00 €
Chemical Identifiers
CAS | 7789-00-6 |
---|---|
Molecular Formula | CrK2O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00011368 |
InChI Key | XMXNVYPJWBTAHN-UHFFFAOYSA-N |
Synonym | potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 |
PubChem CID | 24597 |
ChEBI | CHEBI:75249 |
IUPAC Name | dipotassium;dioxido(dioxo)chromium |
SMILES | [K+].[K+].[O-][Cr]([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12357193
|
Thermo Scientific Acros
447201000 |
100 g | Glass bottle |
33.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12367193
|
Thermo Scientific Acros
447205000 |
500 g | Glass bottle |
106.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-00-6 | |
194.19 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
24597 | |
dipotassium;dioxido(dioxo)chromium |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
CHEBI:75249 | |
[K+].[K+].[O-][Cr]([O-])(=O)=O |
Specifications
975.0°C | |
CrK2O4 | |
MFCD00011368 | |
potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687 | |
XMXNVYPJWBTAHN-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)chromium | |
24597 | |
194.2 | |
Potassium chromate |
7789-00-6 | |
K2CrO4 | |
15,7743 | |
Solubility in water: 640g/L (20°C). Other solubilities: insoluble in alcohol | |
[K+].[K+].[O-][Cr]([O-])(=O)=O | |
194.19 | |
CHEBI:75249 | |
ACS Reagent |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes skin irritation.
May cause respiratory irritation.
May cause cancer by inhalation.
May cause genetic defects.
Causes serious eye irritation.
Very toxic
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-140-5
- Potassium chromate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H350i-May cause cancer by inhalation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only