Learn More
Potassium chloride, ultra dry, 99.95% (metals basis), Thermo Scientific Chemicals
87.50 € - 1246.00 €
Chemical Identifiers
CAS | 7447-40-7 |
---|---|
Molecular Formula | ClK |
Molecular Weight (g/mol) | 74.55 |
MDL Number | MFCD00011360 |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
PubChem CID | 4873 |
ChEBI | CHEBI:32588 |
IUPAC Name | potassium;chloride |
SMILES | [Cl-].[K+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362418
|
Thermo Scientific Alfa Aesar
014466.09 |
10 g | |||||||
11352418
|
Thermo Scientific Alfa Aesar
014466.18 |
50 g | |||||||
11480460
|
Thermo Scientific Alfa Aesar
014466.30 |
250 g |
1246.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher, and to treat hypokalemia (a condition in which the level of potassium will be low in blood) with its pharmaceutical preparation. Furthermore, it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7447-40-7 | |
74.55 | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
4873 | |
potassium;chloride |
ClK | |
MFCD00011360 | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
CHEBI:32588 | |
[Cl-].[K+] |
Specifications
1500°C (sublimation) | |
7447-40-7 | |
Beads | |
ClK | |
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium;chloride | |
4873 | |
74.55 | |
1.49 | |
White beads | |
Potassium chloride |
773°C | |
White | |
99.95% (metals basis) | |
MFCD00011360 | |
Freely soluble in water. Slightly soluble in alcohol. Insoluble in ether. | |
[Cl-].[K+] | |
74.55 | |
CHEBI:32588 | |
Odorless | |
Hygroscopic | |
1.984 |
Safety and Handling
H303: May be harmful if swallowed.
missing translation for 'einecsNumber' : 231-211-8
missing translation for 'rtecsNumber' : TS8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only