missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride 3M/l + AgCl, Chem Lab™
Click to view available options
Quantity:
250 mL
Description
- Molecular Formula: KCl-H20 AgCl saturated
- Molar Mass: 74.56g/mol
- CAS: 7447-40-7
Specifications
Specifications
CAS | 7447-40-7 |
Color | Colorless |
Physical Form | Liquid |
Quantity | 250 mL |
Molecular Formula | ClK |
MDL Number | MFCD00011360 |
Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
SMILES | [Cl-].[K+] |
IUPAC Name | potassium chloride |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction