missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate sesquihydrate, 98.5%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10235321
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 891.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 98.5%
Merck Index 15, 7740
Solubility Information Solubility in water: soluble in water., insoluble in alcohol
Formula Weight 165.24
Percent Purity 98.5%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Potassium carbonate sesquihydrate, For analysis ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium carbonate sesquihydrate, reagent ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'