missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, ReagentPlus™, Honeywell™

Product Code. 15693160
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
12 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Quantity 5 kg
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
IUPAC Name dipotassium;carbonate
Formula Weight 138.21g/mol
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material Potassium Carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'