missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous, Certified AR, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™

Product Code. 10191480
Click to view available options
Quantity:
250 g
500 g
2.5 kg

CK2O3, CAS Number-584-08-7, carbonate of potash, carbonic acid, dipotassium salt, dipotassium carbonate, kaliumcarbonat, pearl ash, potash, potassiumcarbonate, potassium carbonate, potassium carbonate, anhydrous, salt of tartar, 500g, White, 138.21g/mol, BWHMMNNQKKPAPP-UHFFFAOYSA-L

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 2.5 kg
IUPAC Name dipotassium;carbonate
Formula Weight 138.21g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Potassium carbonate anhydrous
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.