Learn More
Potassium bromide, ultra dry, 99.9% (metals basis), Thermo Scientific Chemicals
88.84 € - 911.54 €
Chemical Identifiers
CAS | 7758-02-3 |
---|---|
Molecular Formula | BrK |
Molecular Weight (g/mol) | 119.00 |
MDL Number | MFCD00011358 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
PubChem CID | 253877 |
ChEBI | CHEBI:32030 |
SMILES | [K+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327008
|
Thermo Scientific Alfa Aesar
014700.14 |
25 g |
88.84 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337008
|
Thermo Scientific Alfa Aesar
014700.22 |
100 g |
263.31 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317008
|
Thermo Scientific Alfa Aesar
014700.36 |
500 g |
911.54 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potassium bromide is widely used as an anticonvulsant and a sedative. Also used in the preparation of stable Pd nanocubes that si used in immunolabeling. Potassium bromide is used for the purification of Sarcocystis neurona sporocysts by discontinuous density gradient centrifugation. May be used for removal of peripheral membrane proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
[K+].[Br-] |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 |
Specifications
1435°C | |
7758-02-3 | |
Beads | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium;bromide | |
253877 | |
119.01 | |
Odorless | |
1.559 | |
2.75 g/mL | |
Potassium bromide, Ultra dry |
730°C | |
White | |
BrK | |
14,7618 | |
It is freely soluble in water. very slightly soluble in diethyl ether | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
99.9% | |
Ampule | |
Hygroscopic | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
missing translation for 'einecsNumber' : 231-830-3
missing translation for 'rtecsNumber' : TS7650000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.