Learn More
Potassium bromide, 99+%, ACS reagent, Thermo Scientific Chemicals
57.59 € - 168.84 €
Chemical Identifiers
CAS | 7758-02-3 |
---|---|
Molecular Formula | BrK |
Molecular Weight (g/mol) | 119.00 |
MDL Number | MFCD00011358 |
InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
Synonym | potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 |
PubChem CID | 253877 |
ChEBI | CHEBI:32030 |
IUPAC Name | potassium bromide |
SMILES | [K+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10269750
|
Thermo Scientific Acros
424075000 |
500 g | Plastic bottle |
57.59 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10668144
|
Thermo Scientific Acros
424070025 |
2.5 kg | Plastic bottle |
168.84 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7758-02-3 | |
119.00 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
253877 | |
potassium bromide |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
CHEBI:32030 | |
[K+].[Br-] |
Specifications
1435°C | |
7758-02-3 | |
5.0 to 8.8 (5% soln. at 25°C) | |
99+% | |
KBr | |
15, 7739 | |
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol | |
[K+].[Br-] | |
119.00 | |
CHEBI:32030 | |
≥99% | |
<0.01mmHg at 20°C | |
Potassium bromide |
734°C | |
White | |
Crystalline Solid, Powder, Crystals and/or Chunks | |
BrK | |
MFCD00011358 | |
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
potassium bromide | |
253877 | |
119 | |
ACS Reagent | |
>800°C |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-830-3
missing translation for 'rtecsNumber' : TS7650000
missing translation for 'tsca' : TSCA
- Potassium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Potassium bromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.