missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 98%, extra pure, Thermo Scientific Chemicals

50.85 € - 164.82 €

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10163830
View Documents
Thermo Scientific Acros
196480010
1 kg Plastic bottle
50.85 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10722135
View Documents
Thermo Scientific Acros
196480050
5 kg Plastic bottle
164.82 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
[K+].[Br-]
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030

Specifications

1435.0°C
7758-02-3
Crystalline Powder and/or Chunks
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119
Extra Pure
Potassium bromide, Extra Pure
734.0°C
White
97.5% min. (on dry substance) (Argentometry)
KBr
15, 7739
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
[K+].[Br-]
119.00
CHEBI:32030
98%
1% max.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 231-830-3

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only