Learn More
Potassium bromate, 99.5%, for analysis, Thermo Scientific Chemicals
82.20 € - 173.00 €
Chemical Identifiers
CAS | 7758-01-2 |
---|---|
Molecular Formula | BrKO3 |
Molecular Weight (g/mol) | 167.01 |
MDL Number | MFCD00011359 |
InChI Key | OCATYIAKPYKMPG-UHFFFAOYSA-M |
Synonym | potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat |
PubChem CID | 23673461 |
ChEBI | CHEBI:38211 |
IUPAC Name | potassium;bromate |
SMILES | [O-]Br(=O)=O.[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10043230
|
Thermo Scientific Acros
268392500 |
250 g | Plastic bottle |
82.20 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10227310
|
Thermo Scientific Acros
268390010 |
1 kg | Plastic bottle |
173.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagentChemical Identifiers
7758-01-2 | |
167.01 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
BrKO3 | |
MFCD00011359 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
Specifications
350.0°C | |
White | |
Crystalline Powder | |
BrKO3 | |
MFCD00011359 | |
15, 7738 | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water | |
[O-]Br(=O)=O.[K+] | |
167.01 | |
CHEBI:38211 | |
99.5% | |
0.1% max. (105°C) | |
3.2700g/mL |
7758-01-2 | |
5 to 9 (5% soln. at 20°C) | |
99.5% min. (Iodometry) | |
KBrO3 | |
11,431 | |
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01 | |
Analytical | |
3.27 | |
Potassium bromate, p.a. |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-829-8
missing translation for 'rtecsNumber' : EF8725000
missing translation for 'tsca' : TSCA
RUO – Research Use Only