missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for 3130/3130xl Genetic Analyzers

POP-7™ Polymer is a separation matrix for performing DNA sequencing and fragment analysis on the Applied Biosystems 3130 and the Applied Biosystems 3130xl Genetic Analyzers.

Specifications

For Use With (Application) Fragment Analysis, Sequencing
Product Line POP-7™
View More Specs

Description

Description

  • Easy to use: Pre-formulated, which not only saves time but also saves reproducibility by ensuring consistent formulation
  • Robust formulation: Formulated for sequencing and fragment analysis applications
  • Replenishable: Allow capillaries to be used multiple times

Order Info

Each

Specifications

Specifications

Fragment Analysis, Sequencing
POP-7™
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Refrigerate at 2°C to 8°C.

SDS
Documents

Documents

Product Certifications

For research use only