missing translation for 'onlineSavingsMsg'
Learn More

Polyvinylpyrrolidone, average M.W. 50.000, K30, Thermo Scientific Chemicals

Product Code. 10697323
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Pail
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Color White
pH 3.0 to 5.0 (5% aq. soln.)
Physical Form Crystalline Powder or Crystals
Melting Point 225.0°C
Merck Index 15, 7814
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, practically insoluble in ether
Quantity 2.5 kg
Percent Purity 11.5 to 12.8% (Nitrogen, on anh. subst.) 10ppm 1-vinylpyrrolidin-2-one (HPLC) max. 3% max. 2-pyrrolidone (HPLC)
Identification B,C,D,E comply
Infrared Spectrum Authentic
Packaging Plastic Pail
Viscosity 29 to 32 K−value (1% solid w/v aq. soln.)
Water 5% Max. (K.F.)
Chemical Name or Material Polyvinylpyrrolidone, K29-32
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.