missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl acetate, M.W. ca 50,000, Thermo Scientific™

20.95 € - 80.79 €

Chemical Identifiers

CAS 9003-20-7
Molecular Formula (C4H6O2)n
Molecular Weight (g/mol) 86.09
MDL Number MFCD00084457
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC(-*)C-*
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400683
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12732.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11410683
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12732.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

9003-20-7
86.09
XTXRWKRVRITETP-UHFFFAOYSA-N
7904
ethenyl acetate
(C4H6O2)n
MFCD00084457
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
CHEBI:46916
CC(=O)OC(-*)C-*

Specifications

9003-20-7
MFCD00084457
Soluble in ketones,ethers and aromatic hydrocarbons.
CC(=O)OC(-*)C-*
86.09
CHEBI:46916
Polyvinyl acetate
(C4H6O2)n
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
XTXRWKRVRITETP-UHFFFAOYSA-N
ethenyl acetate
7904
≈50,000
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications