Learn More
TPP™ Polystyrene Petri Dishes
Manufactured from crystal clear polystyrene to provide visual control of cells in stacked dishes using transmitted light. TPP™ Polystyrene Petri Dishes are designed with a gripping ring for easy handling and to prevent accidental dropping of dish bottom.
Specifications
Color | Clear |
---|---|
Material | Polystyrene |
Shape | Round |
Sterility | Sterile |
Type | Petri Dish |
Description
Manufactured from crystal clear polystyrene to provide visual control of cells in stacked dishes using transmitted light. TPP™ Polystyrene Petri Dishes are designed with a gripping ring for easy handling and to prevent accidental dropping of dish bottom.
- Surface treatment of the growth area optimally enhances proliferation of cells
- Serrations in the gripping ring render the dish grip secure
- Yellow marking area on side of the lid, frosted on side of the bottom
- Stacking ring on the lid and corresponding ridge on the base provide secure stacking
- Consistent growth conditions:
- Vents provide airflow between stacked dishes for consistent growth and prevention of condensation
- Cams on the inside of the dish lid function as spacer, limit condensation and enable a constant movement of air
- Numeric "clock" markings (12, 3, 6, 9) in the base of the dish divide it into quadrants for quick orientation
Specifications
Clear | |
Round | |
Petri Dish |
Polystyrene | |
Sterile | |
21 mm |