Learn More
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths
Use these cuvettes for more constant sample temperatures during photometric measurements. Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths feature excellent optical transmission range and small wall thickness for increased heat transfer.
Brand:  Fisherbrand™ FB55923
Product Code. 11817922
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Cuvets with improved photometric properties.
  • Very low variation of extinction values
  • Excellent optical transmission range
Specifications
Specifications
Macro Cuvette
4 mL
100/Pk.
45 mm
Reusable
Plastic
220 to 900 nm
12.5 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Polystyrene Macro Cuvettes for UV and Visible Wavelengths > Capacity: 4mL; Wavelength: 220 to 900nm; QTY: 100/Pk.