Learn More
Sampling Systems™ SteriWare™ Disposable PowderSafe
BIOPHARMA
Powder Spatula complete with a see through, click-on container. The container slips over the blade of the spatula and is securely locked into place. At the laboratory the container can be easily removed.
229.00 €
Specifications
Product Type Powder Sampler
Material Polystyrene
Disposable Single-use
Color White Spatula, Transparent Cover
Certifications/Compliance Conforms to FDA CFR 177.1640, Conforms to EU Regulation 10/2011
View More Specs
Products 1
Product Code Brand Sterility Price Quantity & Availability  
Product Code Brand Sterility Price Quantity & Availability  
11975846
BIOPHARMA
View Documents
Sampling Systems™
8097A-150
Non-sterile
229.00 €
Pack of 100
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
Key Features:
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Supplied with see-through container that securely clips over the spatula
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable device, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Specifications
Specifications
Powder Sampler
Single-use
Conforms to FDA CFR 177.1640, Conforms to EU Regulation 10/2011
230 mm
Polystyrene
White Spatula, Transparent Cover
For Easy, Controlled Powder Discharge