missing translation for 'onlineSavingsMsg'
Learn More

3M™ White Disposable Protective Coveralls, 4515 Series

Product Code. 12942275
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Height (English):
64 to 67 in.
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Height (Metric):
164 to 170 cm
167 to 176 cm
174 to 181 cm
179 to 187 cm
194 to 200 cm
‎186 to 194 cm
‎200 to 206 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Coveralls provide protection from hazardous dusts such as asbestos fibers. Meets PPE Directive Category III, Type 5/6 protection.

  • Breathable material reduces heat build-up and discomfort
  • Excellent barrier against dry particles and limited liquid splash (CE Type 5/6)
  • Robust, highly breathable 50gsm SMS material
  • Meets inward leakage requirements of EN 1073-2 for protection against nuclear particles
  • Two-way zipper with fully sealable storm flap
  • Two-panel hood design
  • Elasticated wrists, waist, and ankles for increased comfort and freedom of movement
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material SMS Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Hood Type 2-panel
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Protection Class PPE Directive Category III, Type 5/6
Model 4515W
Size (English) Chest 49 to 52 in.
Size (Metric) Chest 124 to 132 cm
Height (English) 76 to 78 in.
Height (Metric) 194 to 200 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'