missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Polypropylene Liquid Funnels
These liquid funnels are designed for long life and can be cleaned and re-used.
3.90 € - 22.25 €
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
15869411
|
Sampling Systems™
8022P-100 |
100mm |
3.90 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
15879411
|
Sampling Systems™
8022P-200 |
200mm |
22.25 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
- Crevice-free
- Chemically resistant
- Available in two different top diameters, 100 and 200mm
Specifications
Polypropylene | |
Natural |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title