missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ XTR Adsorbent

Use Chromabond™ XTR with Kieselguhr phase for liquid-liquid extraction. Chromabond™ XTR Adsorbent provides options for Chromabond™ XTR columns.

139.00 € - 1060.00 €

Specifications

Product Type XTR Adsorbent
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10761324
View Documents
Macherey-Nagel™
730595.1000
1000 g
269.00 €
1000g
In Stock
to see stock.
Add to basket
 
10377971
View Documents
Macherey-Nagel™
730595.5000
5000 g
1060.00 €
5kg
In Stock
to see stock.
Add to basket
 
10032841
View Documents
Macherey-Nagel™
730595.500
500 g
139.00 €
500g
In Stock
to see stock.
Add to basket
 
12491498
View Documents
Macherey-Nagel™
730586
50 Pack
289.00 €
Pack of 50
In Stock
to see stock.
Add to basket
 
12481498
View Documents
Macherey-Nagel™
730585
100 pack
289.00 €
Pack of 50
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

Features
  • Base material coarse-grained kieselguhr (also known as diatomaceous earth, hydromatrix, celite)
  • large pore size, high pore volume, constantly high batch-to-batch quality
  • pH working range 1–13
Application
  • Liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel
  • High water loadability without breakthrough of water during elution with organic solvents
  • Also suited for removing small amounts of water from solvents which are not miscible with water
Advantages
  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation ∙ no formation of emulsions
  • High recovery rates
  • Saving of time and solvents
  • Organic solutions need not to be dried after separation
Solvents applicable for elution
  • diethyl ether
  • tert-butyl methyl ether
  • ethyl acetate
  • n-hexane
  • cyclohexane
  • toluene
  • dichloromethane (methylene chloride)
  • trichloromethane (chloroform)
  • solvent mixtures:
  • trichloromethane – methanol (90:10, v/v)
  • trichloromethane – methanol (85:15, v/v)
  • diethyl ether – ethanol (90:10, v/v)
  • diethyl ether – ethanol (80:20, v/v)
  • dichloromethane – 2-propanol (90:10, v/v)
  • dichloromethane – 2-propanol (85:15, v/v)
Specifications

Specifications

XTR Adsorbent
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.