missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semadeni™ Color-Coded Plastic Beakers
Excellent for the storage, mixing or pouring of liquids in the laboratory. Semadeni™ Color-Coded Plastic Beakers are manufactured from durable polypropylene.
Brand: Semadeni™ 8611
This item is not returnable.
View return policy
Description
- Translucent colors
- 30mL capacity
- Graduated
- Optional LDPE lids available for purchase
Specifications
Yellow | |
Polypropylene | |
Beaker | |
Yes |
30 mL | |
Suitable as dosing or laboratory beakers | |
Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction