missing translation for 'onlineSavingsMsg'
Learn More

Polymyxin B Sulfate MP Biomedicals

Product Code. 11487660
Click to view available options
Quantity:
10 mU
25 mU
5 mU
50 mU
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1405-20-5
Molecular Formula C48H84N16O17S
Molecular Weight (g/mol) 1189.36
MDL Number MFCD00131991
InChI Key HNDFYNOVSOOGDU-UHFFFAOYNA-N
Synonym Aerosporin
PubChem CID 133109994
IUPAC Name N-[(2S)-4-amino-1-[[(2S,3R)-1-[[(2S)-4-amino-1-oxo-1-[[(3S,6S,18R)-6,9,18-tris(2-aminoethyl)-15-benzyl-3-[(1R)-1-hydroxyethyl]-12-(2-methylpropyl)-2,5,8,11,14,17,20-heptaoxo-1,4,7,10,13,16,19-heptazacyclotricos-21-yl]amino]butan-2-yl]amino]-3-hydroxy-1-ox
SMILES OS(O)(=O)=O.CC(C)CC1NC(=O)C(CC2=CC=CC=C2)NC(=O)C(CCN)NC(=O)C(CCNC(=O)C(CCO)NC(=O)C(CCN)NC(=O)C(CCN)NC1=O)NC(=O)C(CCN)NC(=O)C(CCO)NC(=O)C(CCN)NC=O

Specifications

Type Polymixin B Sulfate
Physical Form Powder
Formula Weight 1385.61
Color White
Quantity 25 mU
Chemical Name or Material Polymyxin B Sulfate, Aerosporin Activity: >6, 000 u/mg
compliance-icons
Product Identifier
  • Polymyxin B Sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'